Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138403.4 | NP_612412.2 | 6 | 700 | ||
XM_017012793.1 | XP_016868282.1 | 5 | 420 | |||
GenBank mRNA | BC002778.2 | - | 6 | 700 | 3616529 |