Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267895.1 | NP_001254824.1 | 14 | 1275 | ||
NM_133455.3 | NP_597712.2 | 14 | 1281 | |||
XM_011529869.2 | XP_011528171.1 | 15 | 1286 | |||
XM_011529873.2 | XP_011528175.1 | 15 | 1287 | |||
GenBank mRNA | AJ416090.1 | - | 14 | 1148 | ||
AK097163.1 | - | 1 | 456 | |||
BC046358.1 | - | 14 | 1229 | 5770546 | ||
CR456407.1 | - | 14 | 1164 |