Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320845.1 | NP_001307774.1 | 5 | 576 | ||
NM_001320846.1 | NP_001307775.1 | 4 | 474 | |||
NM_001320847.1 | NP_001307776.1 | 4 | 457 | |||
NM_133491.4 | NP_597998.1 | 5 | 559 | |||
XM_017024073.1 | XP_016879562.1 | 5 | 576 | |||
XM_017024074.1 | XP_016879563.1 | 4 | 476 | |||
GenBank mRNA | AF348524.1 | - | 5 | 318 | ||
BC011751.2 | - | 5 | 529 | 3534187 | ||
BC065922.1 | - | 1 | 1017 | |||
BC095466.1 | - | 5 | 514 | |||
BG748008.1 | - | 4 | 384 | |||
BP317593.1 | - | 4 | 475 | |||
HY155008.1 | - | 4 | 460 |