Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282356.1 | NP_001269285.1 | 9 | 1271 | ||
NM_001282357.1 | NP_001269286.1 | 8 | 1108 | |||
NM_052832.3 | NP_439897.1 | 8 | 1232 | |||
NM_134266.1 | NP_599028.1 | 8 | 1232 | |||
GenBank mRNA | AF331521.1 | - | 8 | 1202 | ||
AJ413228.1 | - | 8 | 1201 | |||
AJ413229.2 | - | 8 | 1233 | |||
AJ413230.1 | - | 8 | 1201 | |||
BC113866.1 | - | 7 | 1084 | 40034306 | ||
BC114474.1 | - | 7 | 1072 | 40034303 |