Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTSSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138288.3 | NP_612145.2 | 2 | 758 | ||
GenBank mRNA | AK001993.1 | - | 2 | 739 | ||
AL080066.1 | - | 2 | 739 | |||
BC016805.1 | - | 2 | 740 | 4075637 | ||
BC016808.1 | - | 2 | 740 | 4077762 | ||
BC021701.1 | - | 2 | 739 | 4069530 | ||
BC068480.1 | - | 2 | 852 | 5294549 |