Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138295.3 | NP_612152.1 | 9 | 1290 | ||
XM_017011798.1 | XP_016867287.1 | 10 | 7695 | |||
GenBank mRNA | AB061683.1 | - | 9 | 1256 |