Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322468.1 | NP_001309397.1 | 20989 | |||
NM_004532.5 | NP_004523.3 | 15 | 2263 | |||
NM_018406.6 | NP_060876.5 | 14971 | ||||
NM_138297.4 | NP_612154.2 | 14 | 2110 | |||
GenBank mRNA | AJ010901.1 | - | 15 | 2047 | ||
AJ242541.1 | - | 16 | 2272 | |||
AJ242542.1 | - | 14 | 1972 | |||
AJ242543.1 | - | 15 | 1971 | |||
AJ242544.1 | - | 15 | 1943 | |||
AJ242545.1 | - | 14 | 1882 | |||
AJ242546.1 | - | 13 | 1617 | |||
AJ242547.1 | - | 15 | 2047 | |||
AJ242548.1 | - | 11 | 1369 | |||
AJ242549.1 | - | 15 | 2147 | |||
AJ242550.1 | - | 14 | 1994 | |||
AJ276359.1 | - | 17 | 5157 | |||
AJ277412.1 | - | 16 | 5001 | |||
AJ277505.1 | - | 15 | 4898 | |||
AJ400633.1 | - | 17 | 5171 | |||
AJ400849.1 | - | 17 | 5081 | |||
AJ400850.1 | - | 17 | 5081 | |||
AJ400851.1 | - | 17 | 5171 | |||
AJ400852.1 | - | 16 | 4925 | |||
AJ400853.1 | - | 16 | 4897 | |||
AJ400854.1 | - | 16 | 5002 | |||
AJ400855.1 | - | 16 | 4973 | |||
AJ400856.1 | - | 17 | 5128 | |||
AJ400857.1 | - | 17 | 5157 | |||
AJ400858.1 | - | 17 | 5157 |