Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACMSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307983.1 | NP_001294912.1 | 8, 9 | 981 | ||
NM_138326.2 | NP_612199.2 | 8, 9 | 924 | |||
XM_005263586.4 | XP_005263643.1 | 6, 7 | 1074 | |||
XM_005263588.4 | XP_005263645.1 | 7, 8 | 796 | |||
XM_005263589.4 | XP_005263646.1 | 9, 10 | 1332 | |||
XM_005263590.4 | XP_005263647.1 | 5, 6 | 1638 | |||
XM_011510592.2 | XP_011508894.1 | 8, 9 | 1282 | |||
XM_017003325.1 | XP_016858814.1 | 8, 9 | 1279 | |||
XM_017003326.1 | XP_016858815.1 | 8, 9 | 1929 | |||
GenBank mRNA | AB071418.1 | - | 8, 9 | 857 | ||
BC016018.1 | - | 8, 9 | 983 | 4714682 | ||
BC107420.1 | - | 8, 9 | 864 | 4594027 | ||
KU178859.1 | - | 5, 6 | 683 | |||
KU178860.1 | - | 5, 6 | 618 |