Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138340.4 | NP_612213.2 | 5 | 711 | ||
XM_011525847.2 | XP_011524149.1 | 5 | 684 | |||
XM_017025571.1 | XP_016881060.1 | 5 | 687 | |||
XM_017025572.1 | XP_016881061.1 | 4 | 515 | |||
XM_017025573.1 | XP_016881062.1 | 6 | 496 | |||
XM_017025574.1 | XP_016881063.1 | 4 | 439 | |||
GenBank mRNA | AF007152.1 | - | 5 | 670 | ||
AK024880.1 | - | 5 | 585 | |||
AK298728.1 | - | 5 | 610 | |||
BC021196.2 | - | 5 | 671 | 3942164 |