Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138376.2 | NP_612385.2 | 9, 10 | 1263 | ||
GenBank mRNA | AK074553.1 | - | 9, 10 | 1246 | ||
BC008647.1 | - | 9, 10 | 1204 | |||
BC030822.1 | - | 9, 10 | 1207 | 4243419 | ||
BC053538.1 | - | 9, 10 | 1211 | 5225152 | ||
BX248066.1 | - | 9, 10 | 1035 | |||
BX640732.1 | - | 9, 10 | 1142 | |||
BX648584.1 | - | 8, 9 | 4397 |