Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142650.1 | NP_001136122.1 | 4 | 588 | ||
NM_138394.3 | NP_612403.2 | 3 | 844 | |||
XM_005264639.3 | XP_005264696.2 | 3 | 841 | |||
XM_005264640.3 | XP_005264697.2 | 4 | 587 | |||
XM_011533165.2 | XP_011531467.1 | 3 | 841 | |||
XM_011533166.2 | XP_011531468.1 | 3 | 841 | |||
GenBank mRNA | AK000155.1 | - | 4 | 492 | ||
AK291462.1 | - | 3 | 709 | |||
AK292259.1 | - | 4 | 581 | |||
AL512692.1 | - | 2 | 238 | |||
AY236962.1 | - | 3 | 673 | |||
AY236963.2 | - | 4 | 433 | |||
BC008217.1 | - | 4 | 481 | 3449607 | ||
BC017480.1 | - | 3 | 690 | 3502860 | ||
DA760455.1 | - | 5 | 524 | |||
KU178839.1 | - | 3 | 434 | |||
KU178840.1 | - | 3 | 434 | |||
KU178841.1 | - | 3 | 434 |