Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138431.2 | NP_612440.1 | 1 | 964 | ||
NR_130120.1 | 1 | 964 | ||||
XM_011516806.2 | XP_011515108.1 | 1 | 1919 | |||
XM_017013005.1 | XP_016868494.1 | 1 | 1919 | |||
GenBank mRNA | BC011982.1 | - | 1 | 831 | 3860434 | |
BF688676.1 | - | 1 | 239 |