Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006572.5 | NP_006563.2 | 1 | 313 | ||
XM_011524202.1 | XP_011522504.1 | 1 | 107 | |||
GenBank mRNA | AF493902.1 | - | 1 | 104 | ||
AK301699.1 | - | 1 | 313 | |||
AK313672.1 | - | 1 | 107 | |||
BC036756.1 | - | 1 | 158 | 5548632 | ||
DC335364.1 | - | 1 | 313 | |||
L22075.1 | - | 1 | 145 |