Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138462.2 | NP_612471.1 | 4, 5 | 589 | ||
XM_005266052.4 | XP_005266109.1 | 4, 5 | 1018 | |||
GenBank mRNA | AJ298882.2 | - | 4, 5 | 597 | ||
AX766496.1 | - | 4, 5 | 597 | |||
BC012948.1 | - | 4, 5 | 552 | 4519078 |