Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BRD8 SIRNA Assays ›
Gene Symbol:
BRD8
Gene Name:
bromodomain containing 8
Gene Aliases:
SMAP, SMAP2, p120
Chromosome Location:
Chr.5: 138139766 - 138178669 on Build GRCh38
UniGene ID:
Hs.519337
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
3675
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BRD8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164326.1
NP_001157798.1
5
310
NM_001300961.1
NP_001287890.1
5
315
NM_001300962.1
NP_001287891.1
4
240
NM_001300966.1
NP_001287895.1
5
310
NM_006696.3
NP_006687.3
5
310
NM_139199.1
NP_631938.1
5
310
XM_005271855.2
XP_005271912.1
5
301
XM_005271856.2
XP_005271913.1
5
301
XM_005271857.2
XP_005271914.1
5
301
XM_005271859.3
XP_005271916.1
5
301
XM_005271860.3
XP_005271917.1
5
301
XM_005271861.3
XP_005271918.1
5
301
XM_011543109.1
XP_011541411.1
5
301
XM_011543110.1
XP_011541412.1
5
287
XM_017008969.1
XP_016864458.1
5
301
XM_017008970.1
XP_016864459.1
5
301
XM_017008971.1
XP_016864460.1
5
301
XM_017008972.1
XP_016864461.1
5
301
XM_017008973.1
XP_016864462.1
5
301
XM_017008974.1
XP_016864463.1
5
301
XM_017008975.1
XP_016864464.1
5
301
XM_017008976.1
XP_016864465.1
5
301
GenBank mRNA
AB209079.1
-
5
256
AF016270.1
-
4
242
AK225928.1
-
5
273
AK289951.1
-
5
270
AK293621.1
-
4
200
AK297756.1
-
5
275
AK310380.1
-
3
149
AK312333.1
-
5
270
AL136823.1
-
5
283
AL704983.1
-
5
310
BC008039.2
-
5
252
3009915
BC008076.2
-
5
252
3009915
BU187982.1
-
6
454
X87613.1
-
4
240
X94234.1
-
4
240
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
signal transduction
cell surface receptor signaling pathway
intracellular receptor signaling pathway
regulation of growth
histone H4 acetylation
histone H2A acetylation
transcription factor activity, sequence-specific DNA binding
thyroid hormone receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline