Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L12 SIRNA Assays ›
Gene Symbol:
BCL2L12
Gene Name:
BCL2 like 12
Gene Aliases:
-
Chromosome Location:
Chr.19: 49665072 - 49673916 on Build GRCh38
UniGene ID:
Hs.289052
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
36761
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040668.1
NP_001035758.1
5
1341
NM_001282516.1
NP_001269445.1
5
1344
NM_001282517.1
NP_001269446.1
4
1201
NM_001282519.1
NP_001269448.1
5
1344
NM_138639.1
NP_619580.1
5
1344
NR_104200.1
4
1201
NR_104201.1
5
1344
NR_104202.1
4
1201
NR_104203.1
4
1201
XM_017027345.1
XP_016882834.1
5
1411
XM_017027346.1
XP_016882835.1
4
1271
GenBank mRNA
BC007724.2
-
5
479
BC104004.1
-
5
917
40024338
BC104005.1
-
5
920
40024342
BC104006.1
-
5
920
40024345
BI159786.1
-
4
367
FJ868798.1
-
5
662
FJ868799.1
-
4
519
FJ868800.1
-
5
662
FJ868801.1
-
4
519
FJ868802.1
-
5
662
FJ868803.1
-
4
519
KR820245.1
-
3
1498
KR820246.1
-
3
1498
KR820247.1
-
3
1498
KR820248.1
-
3
1498
KR820250.1
-
3
1547
KR820251.1
-
3
1547
KR820252.1
-
3
1547
KR820253.1
-
3
1547
KR820255.1
-
4
1279
KR820256.1
-
4
1279
KR820257.1
-
4
1279
KR820259.1
-
4
1328
KR820260.1
-
4
1328
KR820261.1
-
4
1328
KR820262.1
-
4
1328
KR820264.1
-
4
1276
KR820265.1
-
4
1276
KR820267.1
-
4
1325
KR820268.1
-
4
1325
KR820269.1
-
4
1325
KR820271.1
-
4
881
KR820272.1
-
4
881
KR820273.1
-
4
881
KR820275.1
-
4
930
KR820276.1
-
4
930
KR820277.1
-
4
930
KR820279.1
-
5
711
KR820280.1
-
5
711
KR820281.1
-
5
711
KR820282.1
-
5
659
KR820283.1
-
5
659
KR820285.1
-
5
708
KR820286.1
-
5
708
KR820287.1
-
5
708
KR820289.1
-
3
1136
KR820290.1
-
3
1136
KR820291.1
-
3
1136
KR820292.1
-
3
1136
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
positive regulation of transcription from RNA polymerase II promoter
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of cellular senescence
p53 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline