Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYEOV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293291.1 | NP_001280220.1 | 3 | 730 | ||
NM_001293294.1 | NP_001280223.1 | 2 | 826 | |||
NM_001293296.1 | NP_001280225.1 | 2 | 658 | |||
NM_001300923.1 | NP_001287852.1 | 3 | 562 | |||
NM_001300924.1 | NP_001287853.1 | 2 | 467 | |||
NM_138768.3 | NP_620123.2 | 3 | 636 | |||
GenBank mRNA | AJ223366.1 | - | 2 | 804 | ||
AK026148.1 | - | 2 | 638 | |||
AK290121.1 | - | 3 | 712 | |||
BC011815.2 | - | 3 | 594 | 3163949 | ||
BM785815.1 | - | 3 | 388 | |||
BM809741.1 | - | 3 | 687 | |||
BM837864.1 | - | 3 | 619 | |||
DA902417.1 | - | 2 | 447 |