Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000445.1 | - | 2 | 797 | ||
BC032769.1 | - | 2 | 788 | |||
BC053894.1 | - | 2 | 849 | 6144625 | ||
X99682.1 | - | 1 | 41 |