Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286430.1 | NP_001273359.1 | 2 | 205 | ||
NM_001323903.1 | NP_001310832.1 | 2 | 205 | |||
NM_001323904.1 | NP_001310833.1 | 2 | 205 | |||
NM_138792.3 | NP_620147.1 | 2 | 205 | |||
XM_017021911.1 | XP_016877400.1 | 2 | 202 | |||
GenBank mRNA | AK055762.1 | - | 2 | 180 | ||
AK297972.1 | - | 2 | 181 | |||
AY302186.1 | - | 2 | 189 | |||
BC018147.2 | - | 2 | 165 | 3847809 |