Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301834.1 | NP_001288763.1 | 3 | 376 | ||
NM_001301836.1 | NP_001288765.1 | 2 | 373 | |||
NM_001301838.1 | NP_001288767.1 | 2 | 601 | |||
NM_138425.3 | NP_612434.1 | 2 | 505 | |||
GenBank mRNA | AK310266.1 | - | 3 | 536 | ||
AK311912.1 | - | 2 | 303 | |||
BC009925.2 | - | 2 | 282 | 3537206 | ||
BE888740.1 | - | 2 | 292 | |||
BM781587.1 | - | 2 | 505 | |||
BP318750.2 | - | 109 | ||||
CB161655.1 | - | 3 | 376 | |||
DB494917.1 | - | 2 | 373 |