Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138435.2 | NP_612444.2 | 2 | 700 | ||
GenBank mRNA | BC011204.1 | - | 2 | 554 | 3455576 | |
BC031099.1 | - | 2 | 700 | 5296483 |