Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037288.1 | NP_001032365.1 | 4 | 1068 | ||
NM_001288744.1 | NP_001275673.1 | 4 | 1008 | |||
NM_001288745.1 | NP_001275674.1 | 3 | 915 | |||
NM_001288746.1 | NP_001275675.1 | 6 | 1362 | |||
NM_138460.2 | NP_612469.1 | 5 | 1161 | |||
XM_017020953.1 | XP_016876442.1 | 6 | 1147 | |||
XM_017020954.1 | XP_016876443.1 | 6 | 1024 | |||
GenBank mRNA | AF479262.2 | - | 5 | 1162 | ||
AF527413.1 | - | 6 | 1145 | |||
AF527414.1 | - | 5 | 792 | |||
AF527948.2 | - | 3 | 686 | |||
AF527949.1 | - | 4 | 779 | |||
AK094840.1 | - | 4 | 1893 | |||
BC013109.1 | - | 5 | 906 | 4214683 |