Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138572.2 | NP_612639.2 | 7 | 772 | ||
XM_011514296.1 | XP_011512598.1 | 7 | 763 | |||
XM_017010241.1 | XP_016865730.1 | 7 | 763 | |||
XM_017010242.1 | XP_016865731.1 | 7 | 926 | |||
XM_017010243.1 | XP_016865732.1 | 7 | 926 | |||
GenBank mRNA | AF465841.1 | - | 7 | 746 | ||
AK057383.1 | - | 7 | 762 | |||
AL833401.1 | - | 7 | 762 | |||
BC119678.1 | - | 7 | 763 | 40116287 | ||
BC119679.1 | - | 7 | 763 | 40116290 |