Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138777.3 | NP_620132.1 | 3 | 568 | ||
NM_199177.2 | NP_954646.1 | 2 | 232 | |||
XM_005252302.3 | XP_005252359.1 | 4 | 636 | |||
XM_011519183.2 | XP_011517485.1 | 4 | 891 | |||
XM_011519184.2 | XP_011517486.1 | 4 | 401 | |||
XM_011519185.2 | XP_011517487.1 | 4 | 636 | |||
XM_011519186.1 | XP_011517488.1 | 3 | 316 | |||
XM_011519187.2 | XP_011517489.1 | 2 | 356 | |||
XM_017015286.1 | XP_016870775.1 | 4 | 636 | |||
XM_017015287.1 | XP_016870776.1 | 3 | 316 | |||
GenBank mRNA | AK291603.1 | - | 3 | 258 | ||
AK291809.1 | - | 3 | 297 | |||
AK292790.1 | - | 3 | 297 | |||
AK298046.1 | - | 3 | 287 | |||
AK310974.1 | - | 2 | 356 | |||
AK316266.1 | - | 4 | 585 | |||
BC013049.1 | - | 2 | 229 | 3882448 | ||
BC053604.1 | - | 2 | 230 | 6008684 | ||
BF982227.1 | - | 303 |