Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138783.3 | NP_620138.2 | 4 | 1298 | ||
GenBank mRNA | AK131037.1 | - | 4 | 1298 | ||
AK314986.1 | - | 4 | 1196 | |||
AY072704.1 | - | 4 | 1167 | |||
BC014187.2 | - | 4 | 1194 | 4634585 | ||
BC016816.1 | - | 4 | 1218 | 4094670 |