Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256129.1 | NP_001243058.1 | 2 | 366 | ||
NM_138795.3 | NP_620150.1 | 2 | 366 | |||
GenBank mRNA | AB118752.1 | - | 2 | 195 | ||
AK127138.1 | - | 2 | 319 | |||
AK313122.1 | - | 2 | 344 | |||
BC015408.1 | - | 2 | 297 | 4391821 | ||
BG876867.1 | - | 2 | 207 | |||
BM548962.1 | - | 2 | 351 | |||
BP325806.1 | - | 2 | 366 | |||
BX421006.2 | - | 2 | 361 |