Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304993.1 | NP_001291922.1 | 8 | 1048 | ||
NM_138432.3 | NP_612441.1 | 9 | 1170 | |||
XM_005253831.4 | XP_005253888.1 | 8 | 1047 | |||
XM_011537846.2 | XP_011536148.1 | 8 | 1583 | |||
XM_017018763.1 | XP_016874252.1 | 8 | 1047 | |||
GenBank mRNA | AF134473.1 | - | 8 | 994 | ||
BC009849.2 | - | 9 | 1149 | 4040570 | ||
BC091479.1 | - | 8 | 1025 | 30344219 |