Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138435.2 | NP_612444.2 | 4, 5 | 1555 | ||
GenBank mRNA | BC011204.1 | - | 4, 5 | 1409 | 3455576 | |
BC031099.1 | - | 4, 5 | 1555 | 5296483 |