Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135674.1 | NP_001129146.1 | 4 | 1352 | ||
NM_001135675.1 | NP_001129147.1 | 4 | 960 | |||
NM_001135676.1 | NP_001129148.1 | 4 | 645 | |||
NM_138436.3 | NP_612445.2 | 4 | 731 | |||
XM_005273398.4 | XP_005273455.1 | 4 | 1266 | |||
GenBank mRNA | BC013035.2 | - | 4 | 693 | 3535608 | |
BI463786.1 | - | 4 | 650 |