Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138450.5 | NP_612459.1 | 2 | 756 | ||
XM_005266253.3 | XP_005266310.1 | 3 | 665 | |||
GenBank mRNA | AF441378.1 | - | 2 | 756 | ||
AK091249.1 | - | 2 | 567 | |||
BC013150.1 | - | 2 | 535 | 4340470 |