Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138459.3 | NP_612468.1 | 5 | 2113 | ||
GenBank mRNA | AJ420576.1 | - | 1 | 380 | ||
AK025467.1 | - | 1 | 343 | |||
AK125119.1 | - | 5 | 2075 | |||
AK126335.1 | - | 6 | 1827 | |||
AK130093.1 | - | 4 | 1430 | |||
BC013026.2 | - | 5 | 2060 | 3531630 | ||
BC063794.1 | - | 5 | 2067 | 6500832 | ||
BC066910.1 | - | 5 | 2107 | 4830860 | ||
BC110325.1 | - | 5 | 2069 | 5553827 |
Gene Symbol: LOC105370109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130366.1 | - | 4 | 1014 |