Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138573.3 | NP_612640.1 | 6 | 582 | ||
XM_017021937.1 | XP_016877426.1 | 11 | 1181 | |||
XM_017021938.1 | XP_016877427.1 | 10 | 3388 | |||
XM_017021939.1 | XP_016877428.1 | 10 | 950 | |||
XM_017021940.1 | XP_016877429.1 | 9 | 3726 | |||
XM_017021941.1 | XP_016877430.1 | 7 | 689 | |||
XM_017021942.1 | XP_016877431.1 | 7 | 707 | |||
XM_017021943.1 | XP_016877432.1 | 7 | 780 | |||
XM_017021945.1 | XP_016877434.1 | 9 | 3677 | |||
XM_017021946.1 | XP_016877435.1 | 9 | 3677 | |||
XM_017021947.1 | XP_016877436.1 | 9 | 3677 | |||
XM_017021948.1 | XP_016877437.1 | 7 | 3410 | |||
GenBank mRNA | AK307362.1 | - | 6 | 560 | ||
AK311204.1 | - | 12 | 1033 | |||
AK311507.1 | - | 7 | 684 | |||
BC017568.1 | - | 6 | 586 | 4071605 | ||
BX538100.1 | - | 7 | 612 | |||
CR749519.1 | - | 8 | 756 |