Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138691.2 | NP_619636.2 | 9 | 914 | ||
XM_017014256.1 | XP_016869745.1 | 6 | 637 | |||
GenBank mRNA | AF417578.1 | - | 9 | 914 | ||
AK307461.1 | - | 7 | 638 | |||
BC103955.1 | - | 4 | 276 | |||
BC103956.1 | - | 4 | 276 | |||
BC114380.1 | - | 4 | 276 | |||
BC141860.1 | - | 4 | 276 |