Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYEOV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293291.1 | NP_001280220.1 | 3 | 2141 | ||
NM_001293294.1 | NP_001280223.1 | 2 | 2237 | |||
NM_001293296.1 | NP_001280225.1 | 2 | 2069 | |||
NM_001300923.1 | NP_001287852.1 | 3 | 1973 | |||
NM_001300924.1 | NP_001287853.1 | 2 | 1878 | |||
NM_138768.3 | NP_620123.2 | 3 | 2047 | |||
GenBank mRNA | AJ223366.1 | - | 2 | 2215 | ||
AK026148.1 | - | 2 | 2049 | |||
BC011815.2 | - | 3 | 2005 | 3163949 |