Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138777.3 | NP_620132.1 | 3 | 612 | ||
NM_199177.2 | NP_954646.1 | 2 | 276 | |||
XM_005252302.3 | XP_005252359.1 | 4 | 680 | |||
XM_011519183.2 | XP_011517485.1 | 4 | 935 | |||
XM_011519184.2 | XP_011517486.1 | 4 | 445 | |||
XM_011519185.2 | XP_011517487.1 | 4 | 680 | |||
XM_011519186.1 | XP_011517488.1 | 3 | 360 | |||
XM_011519187.2 | XP_011517489.1 | 2 | 400 | |||
XM_017015286.1 | XP_016870775.1 | 4 | 680 | |||
XM_017015287.1 | XP_016870776.1 | 3 | 360 | |||
GenBank mRNA | AK291603.1 | - | 3 | 302 | ||
AK291809.1 | - | 3 | 341 | |||
AK292790.1 | - | 3 | 341 | |||
AK298046.1 | - | 3 | 331 | |||
AK310974.1 | - | 2 | 400 | |||
AK316266.1 | - | 4 | 629 | |||
BC013049.1 | - | 2 | 273 | 3882448 | ||
BC053604.1 | - | 2 | 274 | 6008684 | ||
BF982227.1 | - | 347 |