Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPH7 SIRNA Assays ›
Gene Symbol:
DPH7
Gene Name:
diphthamide biosynthesis 7
Gene Aliases:
C9orf112, RRT2, WDR85
Chromosome Location:
Chr.9: 137554899 - 137578935 on Build GRCh38
UniGene ID:
Hs.292570
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
36951
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPH7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_138778.3
NP_620133.1
2
412
XM_005266123.2
XP_005266180.1
2
402
XM_005266128.2
XP_005266185.1
2
402
XM_005266129.2
XP_005266186.1
2
402
XM_011519188.1
XP_011517490.1
2
401
XM_011519189.1
XP_011517491.1
2
513
XM_011519190.2
XP_011517492.1
2
401
XM_011519191.2
XP_011517493.1
2
401
XM_011519192.2
XP_011517494.1
2
401
XM_011519193.2
XP_011517495.1
2
401
XM_011519194.1
XP_011517496.1
2
401
XM_017015289.1
XP_016870778.1
2
401
XM_017015290.1
XP_016870779.1
2
401
XM_017015291.1
XP_016870780.1
2
237
XM_017015292.1
XP_016870781.1
2
401
XM_017015294.1
XP_016870783.1
2
402
XM_017015295.1
XP_016870784.1
2
401
XM_017015296.1
XP_016870785.1
2
237
XM_017015297.1
XP_016870786.1
2
402
XM_017015298.1
XP_016870787.1
2
402
XM_017015299.1
XP_016870788.1
2
128
XM_017015300.1
XP_016870789.1
2
402
XM_017015303.1
XP_016870792.1
2
401
XM_017015305.1
XP_016870794.1
2
402
XM_017015306.1
XP_016870795.1
2
402
XM_017015307.1
XP_016870796.1
2
402
XM_017015309.1
XP_016870798.1
2
402
GenBank mRNA
AK075115.1
-
2
413
AK310987.1
-
2
513
BC003123.1
-
2
351
3502107
BC017335.2
-
2
371
4642600
Back To Top
More Information
Panther Classification:
Molecular Function -
methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-diphthamide biosynthetic process from peptidyl-histidine
diphthine methylesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline