Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM122C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170779.1 | NP_001164250.1 | 3, 4 | 660 | ||
NM_138819.3 | NP_620174.1 | 3, 4 | 660 | |||
XM_005262382.1 | XP_005262439.1 | 4, 5 | 438 | |||
XM_005262383.2 | XP_005262440.1 | 4, 5 | 438 | |||
XM_005262384.4 | XP_005262441.1 | 4, 5 | 438 | |||
XM_005262386.3 | XP_005262443.1 | 4, 5 | 438 | |||
XM_006724734.3 | XP_006724797.1 | 4, 5 | 526 | |||
XM_006724735.1 | XP_006724798.1 | 4, 5 | 438 | |||
XM_006724736.2 | XP_006724799.1 | 4, 5 | 438 | |||
XM_011531295.1 | XP_011529597.1 | 4, 5 | 438 | |||
XM_011531296.2 | XP_011529598.1 | 4, 5 | 438 | |||
XM_011531297.2 | XP_011529599.1 | 4, 5 | 438 | |||
XM_011531298.2 | XP_011529600.1 | 4, 5 | 438 | |||
XM_017029308.1 | XP_016884797.1 | 4, 5 | 438 | |||
GenBank mRNA | BC017868.1 | - | 3, 4 | 659 | 4699951 | |
BC063491.1 | - | 3, 4 | 600 | 5229041 |