Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139057.3 | NP_620688.2 | 3, 4 | 720 | ||
XM_005254872.3 | XP_005254929.1 | 3, 4 | 1636 | |||
XM_011521312.2 | XP_011519614.1 | 3, 4 | 1636 | |||
XM_017021973.1 | XP_016877462.1 | 3, 4 | 1637 | |||
XM_017021974.1 | XP_016877463.1 | 3, 4 | 1639 | |||
XM_017021975.1 | XP_016877464.1 | 3, 4 | 1637 | |||
XM_017021976.1 | XP_016877465.1 | 2, 3 | 349 | |||
XM_017021977.1 | XP_016877466.1 | 3, 4 | 1636 | |||
XM_017021981.1 | XP_016877470.1 | 3, 4 | 1634 | |||
XM_017021984.1 | XP_016877473.1 | 2, 3 | 295 | |||
GenBank mRNA | AJ315735.1 | - | 3, 4 | 637 | ||
AK131344.1 | - | 2, 3 | 266 | |||
DB279404.1 | - | 2, 3 | 350 |