Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308056.1 | NP_001294985.1 | 4 | 393 | ||
NM_001308057.1 | NP_001294986.1 | 4 | 399 | |||
NM_001308058.1 | NP_001294987.1 | 4 | 393 | |||
NM_001308059.1 | NP_001294988.1 | 5 | 428 | |||
NM_001308061.1 | NP_001294990.1 | 3 | 285 | |||
NM_139164.2 | NP_631903.1 | 4 | 399 | |||
NR_131752.1 | 3 | 285 | ||||
XM_017009044.1 | XP_016864533.1 | 3 | 192 | |||
GenBank mRNA | AF480299.1 | - | 3 | 177 | ||
AK054566.1 | - | 4 | 261 | |||
AK290312.1 | - | 4 | 312 | |||
AK308851.1 | - | 3 | 223 | |||
AL710253.1 | - | 4 | 358 | |||
BC042956.1 | - | 4 | 397 | 5278105 | ||
BP234859.1 | - | 5 | 343 | |||
BP247836.1 | - | 3 | 202 | |||
DB475661.1 | - | 4 | 403 | |||
DT220890.1 | - | 4 | 330 |