Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAET1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243325.1 | NP_001230254.1 | 2 | 273 | ||
NM_001243327.1 | NP_001230256.1 | 2 | 381 | |||
NM_001243328.1 | NP_001230257.1 | 3 | 595 | |||
NM_139165.2 | NP_631904.1 | 2 | 381 | |||
XM_011535475.2 | XP_011533777.1 | 4 | 901 | |||
XM_011535476.2 | XP_011533778.1 | 4 | 915 | |||
XM_011535478.2 | XP_011533780.1 | 4 | 898 | |||
XM_011535479.2 | XP_011533781.1 | 4 | 900 | |||
XM_017010285.1 | XP_016865774.1 | 4 | 900 | |||
XM_017010286.1 | XP_016865775.1 | 4 | 900 | |||
XM_017010287.1 | XP_016865776.1 | 4 | 900 | |||
XM_017010288.1 | XP_016865777.1 | 2 | 149 | |||
GenBank mRNA | AY069961.1 | - | 2 | 308 | ||
AY176317.1 | - | 2 | 248 | |||
AY252119.1 | - | 2 | 248 | |||
AY359075.1 | - | 3 | 595 | |||
EF489426.1 | - | 2 | 248 |