Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SREK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077199.2 | NP_001070667.1 | 5, 6 | 913 | ||
NM_001270492.1 | NP_001257421.1 | 6, 7 | 1037 | |||
NM_001323527.1 | NP_001310456.1 | 6, 7 | 1037 | |||
NM_001323529.1 | NP_001310458.1 | 5, 6 | 913 | |||
NM_001323533.1 | NP_001310462.1 | 5, 6 | 913 | |||
NM_139168.3 | NP_631907.1 | 6, 7 | 1066 | |||
XM_011543171.2 | XP_011541473.1 | 5, 6 | 1138 | |||
XM_017009055.1 | XP_016864544.1 | 5, 6 | 810 | |||
GenBank mRNA | AF459094.1 | - | 5, 6 | 747 | ||
AK091758.1 | - | 5, 6 | 796 | |||
AK308287.1 | - | 6, 7 | 940 | |||
BC047322.1 | - | 5, 6 | 782 | |||
BC067770.1 | - | 6, 7 | 1067 | 4827589 | ||
BC112343.1 | - | 3, 4 | 435 | 40008558 |