Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139178.3 | NP_631917.1 | 3, 4 | 634 | ||
GenBank mRNA | AB042029.1 | - | 3, 4 | 597 | ||
AK315266.1 | - | 3, 4 | 630 | |||
BC067257.1 | - | 3, 4 | 567 | 6176933 | ||
BC103812.1 | - | 3, 4 | 372 | 40000372 | ||
BC103813.1 | - | 3, 4 | 372 | 40000375 | ||
BC103814.2 | - | 3, 4 | 372 | 40000376 |