Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGIF2LY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139214.2 | NP_631960.1 | 2 | 647 | ||
GenBank mRNA | AF332223.1 | - | 2 | 647 | ||
AK292609.1 | - | 2 | 691 | |||
BC128604.1 | - | 1 | 617 | 40003247 |