Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040272.5 | NP_001035362.3 | 3 | 362 | ||
NM_052866.4 | NP_443098.3 | 3 | 362 | |||
XM_011518063.2 | XP_011516365.1 | 5 | 500 | |||
XM_011518064.2 | XP_011516366.1 | 4 | 615 | |||
XM_017015310.1 | XP_016870799.1 | 4 | 7137 | |||
XM_017015311.1 | XP_016870800.1 | 5 | 500 | |||
XM_017015312.1 | XP_016870801.1 | 4 | 616 | |||
XM_017015313.1 | XP_016870802.1 | 4 | 263 | |||
XM_017015314.1 | XP_016870803.1 | 5 | 500 | |||
GenBank mRNA | AF176313.1 | - | 3 | 260 | ||
AK290667.1 | - | 3 | 348 | |||
AK291956.1 | - | 3 | 302 | |||
AY358327.1 | - | 3 | 342 | |||
BC030262.1 | - | 3 | 274 | 5201199 | ||
CX758410.1 | - | 3 | 261 | |||
DA598045.1 | - | 3 | 337 |