Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139286.3 | NP_644815.1 | 4 | 466 | ||
XM_017014573.1 | XP_016870062.1 | 4 | 433 | |||
XM_017014574.1 | XP_016870063.1 | 4 | 898 | |||
XM_017014575.1 | XP_016870064.1 | 4 | 428 | |||
GenBank mRNA | AF503918.1 | - | 1 | 107 | ||
BC042534.1 | - | 4 | 421 | 4751173 | ||
BC066300.1 | - | 4 | 451 | 5258904 | ||
BF244254.1 | - | 4 | 463 | |||
CD685602.1 | - | 4 | 471 |