Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138809.3 | NP_620164.1 | 2 | 583 | ||
XM_017009034.1 | XP_016864523.1 | 2 | 347 | |||
XM_017009035.1 | XP_016864524.1 | 2 | 303 | |||
GenBank mRNA | AJ278125.1 | - | 2 | 376 | ||
AK074197.1 | - | 2 | 374 | |||
AK315390.1 | - | 1 | 137 | |||
BC001573.1 | - | 2 | 289 | 3454199 | ||
BX647141.1 | - | 1 | 80 |