Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012325.2 | NP_036457.1 | 3 | 388 | ||
XM_011528696.2 | XP_011526998.1 | 3 | 390 | |||
GenBank mRNA | AK297273.1 | - | 3 | 237 | ||
AK312590.1 | - | 3 | 305 | |||
BC040410.1 | - | 3 | 315 | |||
BC106068.1 | - | 3 | 318 | 6527006 | ||
BC109281.3 | - | 3 | 312 | 40031641 | ||
BC128442.1 | - | 3 | 283 | |||
BJ992159.1 | - | 3 | 388 | |||
U24166.1 | - | 3 | 313 |