Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM122C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138819.3 | NP_620174.1 | 5, 6 | 748 | ||
XM_005262382.1 | XP_005262439.1 | 6, 7 | 526 | |||
XM_005262384.4 | XP_005262441.1 | 6, 7 | 526 | |||
XM_006724734.3 | XP_006724797.1 | 6, 7 | 614 | |||
XM_006724735.1 | XP_006724798.1 | 6, 7 | 526 | |||
XM_006724736.2 | XP_006724799.1 | 6, 7 | 526 | |||
XM_011531295.1 | XP_011529597.1 | 6, 7 | 526 | |||
XM_011531296.2 | XP_011529598.1 | 6, 7 | 526 | |||
XM_011531297.2 | XP_011529599.1 | 6, 7 | 526 | |||
XM_011531298.2 | XP_011529600.1 | 6, 7 | 526 | |||
XM_011531299.1 | XP_011529601.1 | 3, 4 | 126 | |||
XM_011531300.2 | XP_011529602.1 | 3, 4 | 126 | |||
XM_011531301.2 | XP_011529603.1 | 3, 4 | 121 | |||
XM_011531302.2 | XP_011529604.1 | 3, 4 | 122 | |||
XM_017029308.1 | XP_016884797.1 | 6, 7 | 526 | |||
XM_017029309.1 | XP_016884798.1 | 3, 4 | 203 | |||
GenBank mRNA | AK295485.1 | - | 7, 8 | 1383 | ||
BC017868.1 | - | 5, 6 | 747 | 4699951 |