Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VANGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172412.1 | NP_001165883.1 | 3, 4 | 418 | ||
NM_138959.2 | NP_620409.1 | 3, 4 | 474 | |||
GenBank mRNA | AB057596.1 | - | 3, 4 | 418 | ||
AB075805.1 | - | 2, 3 | 215 | |||
BC032773.1 | - | 1, 2 | 142 | |||
BC065272.1 | - | 3, 4 | 451 | 6165098 | ||
BM552933.1 | - | 493 |