Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL31RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242636.1 | NP_001229565.1 | 3 | 425 | ||
NM_001242637.1 | NP_001229566.1 | 3 | 434 | |||
NM_001242638.1 | NP_001229567.1 | 5 | 798 | |||
NM_001242639.1 | NP_001229568.1 | 4 | 569 | |||
NM_001297570.1 | NP_001284499.1 | 3 | 434 | |||
NM_139017.5 | NP_620586.3 | 3 | 434 | |||
XM_011543142.2 | XP_011541444.1 | 3 | 267 | |||
XM_011543143.2 | XP_011541445.1 | 3 | 457 | |||
GenBank mRNA | AF106913.1 | - | 3 | 298 | ||
AF486620.1 | - | 2 | 146 | |||
AY358117.1 | - | 2 | 146 | |||
AY358740.1 | - | 3 | 434 | |||
AY499339.1 | - | 3 | 307 | |||
AY499340.1 | - | 3 | 307 | |||
AY499341.1 | - | 3 | 307 | |||
AY499342.1 | - | 5 | 681 | |||
BC110490.2 | - | 4 | 361 | 40007489 |